CAS RN 87-55-8
CAS RN 87-55-8
IUPAC Name:
2-methyl-5-oxocyclopent-1-en-1-yl propanoate
Synonyms:
2-methyl-5-oxocyclopent-1-en-1-yl propanoate
Molecular Weight (g/mol):
168.19
Molecular Formula:
C9H12O3
InChi Key:
NKRMCRMCMGRKHS-UHFFFAOYSA-N
SMILES:
CCC(=O)OC1=C(C)CCC1=O
1
–
1
of
1
results
1
2-Methyl-5-oxo-1-cyclopentenyl Propionate, TRC
CAS: 87-55-8 Molecular Formula: C9H12O3 Molecular Weight (g/mol): 168.18 IUPAC Name: (2-methyl-5-oxocyclopenten-1-yl) propanoate SMILES: CCC(=O)OC1=C(C)CCC1=O