accessibility menu, dialog, popup

UserName

OHHOO(R)OH(S)HO(S)HO

CAS RN 87-81-0

IUPAC Name: (3S,4S,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: d-tagatose lyxo-hexulose D-lyxo-2-hexulose D-tagatose lyxo-2-hexulose tagatose, D- D-lyxo-hex-2-ulose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: LKDRXBCSQODPBY-OEXCPVAWSA-N
SMILES: OCC1(O)OC[C@@H](O)[C@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)