accessibility menu, dialog, popup

UserName

OHClOClOOH

CAS RN 87-88-7

IUPAC Name: 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
Synonyms: chloranilic acid
Molecular Weight (g/mol): 208.98
Molecular Formula: C6H2Cl2O4
InChi Key: IPPWILKGXFOXHO-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)