accessibility menu, dialog, popup

UserName

CAS RN 870-63-3

CH3CH3Br

CAS RN 870-63-3

IUPAC Name: 1-bromo-3-methylbut-2-ene
Synonyms: 1-bromo-3-methylbut-2-ene
Molecular Weight (g/mol): 149.03
Molecular Formula: C5H9Br
InChi Key: LOYZVRIHVZEDMW-UHFFFAOYSA-N
SMILES: CC(C)=CCBr
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)