accessibility menu, dialog, popup

UserName

CHCl

CAS RN 873-31-4

IUPAC Name: 1-chloro-2-ethynylbenzene
Synonyms: 1-chloro-2-ethynylbenzene
Molecular Weight (g/mol): 136.58
Molecular Formula: C8H5Cl
InChi Key: DGLHLIWXYSGYBI-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C#C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)