accessibility menu, dialog, popup

UserName

CAS RN 874290-64-9

IUPAC Name: [4-(5-oxopyrazolidin-3-yl)phenyl]boronic acid
Synonyms: 4-(5-oxopyrazolidin-3-yl)phenylboronic acid
Molecular Weight (g/mol): 206.01
Molecular Formula: C9H11BN2O3
InChi Key: INZPXASHKGKJCN-UHFFFAOYNA-N
SMILES: OB(O)C1=CC=C(C=C1)C1CC(=O)NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results