accessibility menu, dialog, popup

UserName

CAS RN 87440-88-8

IUPAC Name: 2-(bromomethyl)pyridin-3-ol hydrobromide
Synonyms: 2-(bromomethyl)pyridin-3-ol hydrobromide
Molecular Weight (g/mol): 268.94
Molecular Formula: C6H7Br2NO
InChi Key: YEZLFBCEZYSGQC-UHFFFAOYSA-N
SMILES: Br.OC1=C(CBr)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results