accessibility menu, dialog, popup

UserName

CAS RN 874839-13-1

IUPAC Name: 4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzene-1-sulfonyl chloride
Synonyms: 4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulfonyl chloride
Molecular Weight (g/mol): 337.7
Molecular Formula: C12H7ClF3NO3S
InChi Key: FDAYBQVWHDJJOL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)N=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)