accessibility menu, dialog, popup

UserName

CAS RN 879896-63-6

OSOOOHBr

CAS RN 879896-63-6

IUPAC Name: 7-bromo-2H,3H-thieno[3,4-b][1,4]dioxine-5-carboxylic acid
Synonyms: 7-bromo-2H,3H-thieno[3,4-b][1,4]dioxine-5-carboxylic acid
Molecular Weight (g/mol): 265.08
Molecular Formula: C7H5BrO4S
InChi Key: KIURUSHZGLZLSQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2OCCOC2=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results