accessibility menu, dialog, popup

UserName

CAS RN 880362-05-0

CH3NNNH

CAS RN 880362-05-0

IUPAC Name: 1-(3-methylpyridin-2-yl)-1,4-diazepane
Synonyms: 1-(3-methylpyridin-2-yl)-1,4-diazepane
Molecular Weight (g/mol): 191.28
Molecular Formula: C11H17N3
InChi Key: ATUSBQCZIPECGZ-UHFFFAOYSA-N
SMILES: CC1=C(N=CC=C1)N1CCCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results