accessibility menu, dialog, popup

UserName

CAS RN 881375-00-4

H3CNNONHSOONO

CAS RN 881375-00-4

IUPAC Name: 2-(4-ethylpiperazin-1-yl)-N-{6-[2-(morpholin-4-yl)-4-oxo-4H-chromen-8-yl]-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-3-yl}acetamide
Synonyms: 2-(4-ethylpiperazin-1-yl)-N-{6-[2-(morpholin-4-yl)-4-oxochromen-8-yl]-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-3-yl}acetamide
Molecular Weight (g/mol): 582.72
Molecular Formula: C33H34N4O4S
InChi Key: AATCBLYHOUOCTO-UHFFFAOYSA-N
SMILES: CCN1CCN(CC(=O)NC2=CC=C(C3=C2C2=C(S3)C=CC=C2)C2=C3OC(=CC(=O)C3=CC=C2)N2CCOCC2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)