accessibility menu, dialog, popup

UserName

CH3NNSOONH2ClCH3

CAS RN 88398-46-3

IUPAC Name: 5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide
Synonyms: 5-chloro-1,3-dimethylpyrazole-4-sulfonamide
Molecular Weight (g/mol): 209.65
Molecular Formula: C5H8ClN3O2S
InChi Key: NYDANSQFEDVGJL-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(=C1Cl)S(N)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results