accessibility menu, dialog, popup

UserName

CAS RN 88398-93-0

IUPAC Name: 5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride
Synonyms: 5-chloro-1,3-dimethylpyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 229.08
Molecular Formula: C5H6Cl2N2O2S
InChi Key: HWGVUNSKAPCFNF-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(=C1Cl)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results