accessibility menu, dialog, popup

UserName

OON

CAS RN 884507-34-0

IUPAC Name: 4-(oxan-4-yloxy)benzonitrile
Synonyms: 4-(oxan-4-yloxy)benzonitrile
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: ITXQSYHCKPGZLV-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results