accessibility menu, dialog, popup

UserName

CAS RN 885268-36-0

IUPAC Name: 2-chloro-N-(5-phenyl-1,2,4-thiadiazol-3-yl)acetamide
Synonyms: 2-chloro-N-(5-phenyl-1,2,4-thiadiazol-3-yl)acetamide
Molecular Weight (g/mol): 253.7
Molecular Formula: C10H8ClN3OS
InChi Key: VUNFVGABWSAHLZ-UHFFFAOYSA-N
SMILES: ClCC(=O)NC1=NSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results