accessibility menu, dialog, popup

UserName

ONNNOBr

CAS RN 885952-23-8

IUPAC Name: 4-[4-(benzyloxy)-5-bromopyrimidin-2-yl]morpholine
Synonyms: 4-[4-(benzyloxy)-5-bromopyrimidin-2-yl]morpholine
Molecular Weight (g/mol): 350.22
Molecular Formula: C15H16BrN3O2
InChi Key: DMIIRAXOFYSJQT-UHFFFAOYSA-N
SMILES: BrC1=CN=C(N=C1OCC1=CC=CC=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results