accessibility menu, dialog, popup

UserName

NONOOH

CAS RN 88598-08-7

IUPAC Name: 1,2,5-oxadiazole-3-carboxylic acid
Synonyms: 1,2,5-oxadiazole-3-carboxylic acid
Molecular Weight (g/mol): 114.06
Molecular Formula: C3H2N2O3
InChi Key: JBLHCUQCDKBPGY-UHFFFAOYSA-N
SMILES: OC(=O)C1=NON=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results