accessibility menu, dialog, popup

UserName

NNClBr

CAS RN 886371-28-4

IUPAC Name: 3-bromo-6-chloroimidazo[1,2-a]pyridine
Synonyms: 3-bromo-6-chloroimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 231.48
Molecular Formula: C7H4BrClN2
InChi Key: LIBKDAUSMLPFBF-UHFFFAOYSA-N
SMILES: ClC1=CN2C(Br)=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results