accessibility menu, dialog, popup

UserName

H3COFOFFF

CAS RN 886501-44-6

IUPAC Name: 1-[4-fluoro-3-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[4-fluoro-3-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: ATZQRDFMSBKRCC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results