accessibility menu, dialog, popup

UserName

N(E/Z)

CAS RN 890-51-7

IUPAC Name: N-(naphthalen-1-yl)-1-phenylmethanimine
Synonyms: N-(naphthalen-1-yl)-1-phenylmethanimine
Molecular Weight (g/mol): 231.3
Molecular Formula: C17H13N
InChi Key: NQPYSYDZNCHIQY-UHFFFAOYSA-N
SMILES: C(=NC1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results