accessibility menu, dialog, popup

UserName

H3CSiH3CCH3OH

CAS RN 89530-34-7

IUPAC Name: 1-phenyl-3-(trimethylsilyl)prop-2-yn-1-ol
Synonyms: 1-phenyl-3-(trimethylsilyl)prop-2-yn-1-ol
Molecular Weight (g/mol): 204.34
Molecular Formula: C12H16OSi
InChi Key: RVTDALNDYLDVMN-UHFFFAOYNA-N
SMILES: C[Si](C)(C)C#CC(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results