CAS RN 895543-25-6
CAS RN 895543-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(2-tert-Butyl-4-bromophenyl)pyrrolidin-2-one, TRC
CAS: 895543-25-6 Molecular Formula: C14H18BrNO Molecular Weight (g/mol): 296.2 Synonym: 1-[4-Bromo-2-(1,1-dimethylethyl)phenyl]-2-pyrrolidinone IUPAC Name: 1-(4-bromo-2-tert-butylphenyl)pyrrolidin-2-one SMILES: BrC1=CC=C(N2CCCC2=O)C(C(C)(C)C)=C1