accessibility menu, dialog, popup

UserName

OOOOH

CAS RN 898289-29-7

IUPAC Name: 2-(oxan-4-yloxy)benzoic acid
Synonyms: 2-(oxan-4-yloxy)benzoic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: AVRDARROMNUMER-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1OC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results