accessibility menu, dialog, popup

UserName

CAS RN 898289-35-5

IUPAC Name: 2-(oxan-4-yloxy)aniline
Synonyms: 2-(oxan-4-yloxy)aniline
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: CLGPZXHLRKSEDD-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1OC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results