accessibility menu, dialog, popup

UserName

CAS RN 90-16-4

ONNHN

CAS RN 90-16-4

IUPAC Name: 1,4-dihydro-1,2,3-benzotriazin-4-one
Synonyms: 1,2,3-benzotriazin-4(1H)-one
Molecular Weight (g/mol): 147.14
Molecular Formula: C7H5N3O
InChi Key: DMSSTTLDFWKBSX-UHFFFAOYSA-N
SMILES: O=C1N=NNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)