CAS RN 9002-24-8
CAS RN 9002-24-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amberlite™ IRA-400(Cl), ion exchange resin, Thermo Scientific™
CAS: 9002-24-8 Molecular Formula: Styrene-DVB; Strongly basic gel type resin MDL Number: MFCD00132708