accessibility menu, dialog, popup

UserName

CAS RN 9003-33-2

OOn

CAS RN 9003-33-2

IUPAC Name: [(ethenyloxy)methoxy]ethene
Synonyms: [(ethenyloxy)methoxy]ethene
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: DFFZYNIDEOOVAU-UHFFFAOYSA-N
SMILES: C=COCOC=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)