accessibility menu, dialog, popup

UserName

CAS RN 9004-36-8

(R)(S)(R)O(R)OO(S)ORgpO(R)ORgpRgp(S)O(R)(R)ORgpORgpORgpn

CAS RN 9004-36-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C12H14O10)n
InChi Key:
SMILES: *-O[C@@H]1[C@@H](CO*)O[C@@H](O[C@@H]2[C@@H](CO*)OC(-*)[C@H](O*)[C@H]2O*)[C@H](O*)[C@H]1O*
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)