accessibility menu, dialog, popup

UserName

CAS RN 9004-95-9

A(a)

CAS RN 9004-95-9

IUPAC Name: 3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxahexaheptacontan-1-ol
Synonyms: ceteth-2 ceteth-17 ceteth-18 ceteth-13 ceteth-14 ceteth-15 ceteth-16 ceteth-24 ceteth-25 ceteth-20
Molecular Weight (g/mol): 1123.51
Molecular Formula: C56H114O21
InChi Key: NLMKTBGFQGKQEV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

 No Refinements