accessibility menu, dialog, popup

UserName

HO(R)OO(S)(R)HO(R)OHOO(S)(R)HOOHO(R)OH(R)OH(R)OH(R)OH(S)HO(S)HO

CAS RN 9008-22-4

IUPAC Name: (3R,4S,5S,6R)-2-{[(2R,3R,4S,5R)-3,5-dihydroxy-2-(hydroxymethyl)-6-{[(3R,4S,5R,6R)-2,3,5-trihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}oxan-4-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: laminarin laminaran
Molecular Weight (g/mol): 504.44
Molecular Formula: C18H32O16
InChi Key: DBTMGCOVALSLOR-VPNXCSTESA-N
SMILES: OC[C@H]1OC(O[C@H]2[C@H](O)[C@@H](CO)OC(O[C@@H]3[C@@H](O)C(O)O[C@H](CO)[C@H]3O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)