accessibility menu, dialog, popup

UserName

CH3NNHOOCH3CH3H3C

CAS RN 90101-22-7

IUPAC Name: tert-butyl N-(6-methylpyridin-2-yl)carbamate
Synonyms: tert-butyl N-(6-methylpyridin-2-yl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: XSVAARVWQDEAEL-UHFFFAOYSA-N
SMILES: CC1=NC(NC(=O)OC(C)(C)C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results