accessibility menu, dialog, popup

UserName

CH3NHOCH3

CAS RN 902836-81-1

IUPAC Name: 2-methyl-2-(4-methylphenyl)morpholine
Synonyms: 2-methyl-2-(4-methylphenyl)morpholine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: NQYMBSUCKZVKFF-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)C1(C)CNCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results