accessibility menu, dialog, popup

UserName

CH3OONH

CAS RN 902837-27-8

IUPAC Name: 3-(2-methoxyphenoxy)piperidine
Synonyms: 3-(2-methoxyphenoxy)piperidine
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: QWVSMVKBNRCNFJ-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results