accessibility menu, dialog, popup

UserName

HO(R)OH(R)OH(S)OH(R)OHO

CAS RN 9050-36-6

IUPAC Name: (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Synonyms: (+)-glucose aldehydo-D-glucose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: GZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)