CAS RN 9057-02-7
CAS RN 9057-02-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
(C18H30O15)AHO
InChi Key:
—
SMILES:
*.O-*.[H][C@]1(-*)O[C@H](CO)[C@@H](O[C@@]2([H])O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO-*)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
1
–
2
of
2
results
1
Pullulan
CAS: 9057-02-7
2
Pullulan, MP Biomedicals™
CAS: 9057-02-7 Molecular Formula: (C18H30O15)AHO InChI Key: GFZFEWWPMNSVBS-WVZDODFGSA-N Synonym: Pullulan from Aureobasidium pullulans PubChem CID: 131636581 IUPAC Name: (2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol SMILES: COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O