accessibility menu, dialog, popup

UserName

H3CNNOClHCl

CAS RN 906352-57-6

IUPAC Name: 1-methyl-1H-1,3-benzodiazole-5-carbonyl chloride hydrochloride
Synonyms: 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride
Molecular Weight (g/mol): 231.08
Molecular Formula: C9H8Cl2N2O
InChi Key: SGNORNZSRSQMEE-UHFFFAOYSA-N
SMILES: Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results