accessibility menu, dialog, popup

UserName

H3COONNN

CAS RN 906352-67-8

IUPAC Name: ethyl 1-(4-cyanopyridin-2-yl)piperidine-4-carboxylate
Synonyms: ethyl 1-(4-cyanopyridin-2-yl)piperidine-4-carboxylate
Molecular Weight (g/mol): 259.31
Molecular Formula: C14H17N3O2
InChi Key: IAIJZMCYSMAKQF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCN(CC1)C1=NC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results