accessibility menu, dialog, popup

UserName

OOS

CAS RN 906352-93-0

IUPAC Name: 4-(thiophen-2-yl)oxane-4-carbaldehyde
Synonyms: 4-(thiophen-2-yl)oxane-4-carbaldehyde
Molecular Weight (g/mol): 196.26
Molecular Formula: C10H12O2S
InChi Key: BMUZFVLYGIZNAH-UHFFFAOYSA-N
SMILES: O=CC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results