accessibility menu, dialog, popup

UserName

CH3NNNH2

CAS RN 910037-08-0

IUPAC Name: 3-(1-methyl-1H-pyrazol-5-yl)aniline
Synonyms: 3-(2-methylpyrazol-3-yl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: KEYWGUASUNFUCS-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C1=CC(N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results