accessibility menu, dialog, popup

UserName

CH3NONHN

CAS RN 912761-48-9

IUPAC Name: 4-(3-ethyl-1,2,4-oxadiazol-5-yl)piperidine
Synonyms: 4-(3-ethyl-1,2,4-oxadiazol-5-yl)piperidine
Molecular Weight (g/mol): 181.24
Molecular Formula: C9H15N3O
InChi Key: PTQDWAPTOMWGGM-UHFFFAOYSA-N
SMILES: CCC1=NOC(=N1)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results