accessibility menu, dialog, popup

UserName

CAS RN 913835-65-1

IUPAC Name: (pyridin-4-yl)boronic acid hydrochloride
Synonyms: pyridin-4-ylboronic acid hydrochloride
Molecular Weight (g/mol): 159.38
Molecular Formula: C5H7BClNO2
InChi Key: NCZXYQJUECNKAD-UHFFFAOYSA-N
SMILES: Cl.OB(O)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results