accessibility menu, dialog, popup

UserName

CAS RN 91419-48-6

IUPAC Name: tert-butyl 4-carbamoylpiperidine-1-carboxylate
Synonyms: tert-butyl 4-carbamoylpiperidine-1-carboxylate
Molecular Weight (g/mol): 228.29
Molecular Formula: C11H20N2O3
InChi Key: YHFUWPUJUMZXBD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results