accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3C

CAS RN 914675-52-8

IUPAC Name: 2-{[1,1'-biphenyl]-2-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-{[1,1'-biphenyl]-2-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 280.17
Molecular Formula: C18H21BO2
InChi Key: WCXWQEUBHZKNMQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results