accessibility menu, dialog, popup

UserName

CAS RN 91510-62-2

IUPAC Name: (2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
Synonyms: (2S,3R,4S,5R,6R)-3,4,5,6-tetrahydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 193.13
Molecular Formula: C6H9O7
InChi Key: AEMOLEFTQBMNLQ-DTEWXJGMSA-M
SMILES: O[C@@H]1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results