accessibility menu, dialog, popup

UserName

CAS RN 916766-85-3

CH3ONHCH3

CAS RN 916766-85-3

IUPAC Name: methyl[(2-methylfuran-3-yl)methyl]amine
Synonyms: methyl[(2-methylfuran-3-yl)methyl]amine
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: IDODRBWAEHXURE-UHFFFAOYSA-N
SMILES: CNCC1=C(C)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results