accessibility menu, dialog, popup

UserName

CH3NNNSBr

CAS RN 919352-66-2

IUPAC Name: 1-(4-bromo-1,3-thiazol-2-yl)-4-methylpiperazine
Synonyms: 1-(4-bromo-1,3-thiazol-2-yl)-4-methylpiperazine
Molecular Weight (g/mol): 262.17
Molecular Formula: C8H12BrN3S
InChi Key: GHLARJPRKBAWMZ-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=NC(Br)=CS1

 No Refinements