accessibility menu, dialog, popup

UserName

CAS RN 921939-08-4

CH3NNFFFOHOO

CAS RN 921939-08-4

IUPAC Name: 1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Synonyms: 1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 286.21
Molecular Formula: C12H9F3N2O3
InChi Key: JLFWKPKAZSVCKI-UHFFFAOYSA-N
SMILES: CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results