accessibility menu, dialog, popup

UserName

CAS RN 92398-47-5

H3COONH2HCl

CAS RN 92398-47-5

IUPAC Name: methyl 1-aminocyclobutane-1-carboxylate hydrochloride
Synonyms: methyl 1-aminocyclobutane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: LLCSDOKIBIMJNU-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1(N)CCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results