CAS RN 926921-76-8
IUPAC Name:
[(2-bromofuran-3-yl)methyl](methyl)amine hydrochloride
Synonyms:
[(2-bromofuran-3-yl)methyl](methyl)amine hydrochloride
Molecular Weight (g/mol):
226.5
Molecular Formula:
C6H9BrClNO
InChi Key:
GYLBKUAMHLATCB-UHFFFAOYSA-N
SMILES:
Cl.CNCC1=C(Br)OC=C1