CAS RN 928-97-2
CAS RN 928-97-2
IUPAC Name:
(3E)-hex-3-en-1-ol
Synonyms:
trans-3-hexenol
Molecular Weight (g/mol):
100.16
Molecular Formula:
C6H12O
InChi Key:
UFLHIIWVXFIJGU-ONEGZZNKSA-N
SMILES:
CC\C=C\CCO
1
–
2
of
2
results
1
trans-3-Hexen-1-ol, 97%
CAS: 928-97-2 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00002960 InChI Key: UFLHIIWVXFIJGU-ONEGZZNKSA-N Synonym: trans-3-hexen-1-ol,e-hex-3-en-1-ol,e-3-hexen-1-ol,trans-3-hexenol,3-hexen-1-ol,3e-hex-3-en-1-ol,3-hexen-1-ol, e,trans-hex-3-en-1-ol,unii-4e0nfr5b3u,3e-hexen-1-ol PubChem CID: 5284503 IUPAC Name: (E)-hex-3-en-1-ol SMILES: CCC=CCCO
2
trans-3-Hexen-1-ol, TRC
CAS: 928-97-2 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.16 Synonym: (E)-3-Hexen-1-ol,trans-3-Hexen-1-ol,(3E)-3-Hexen-1-ol,(3E)-Hex-3-en-1-ol,(E)-3-Hexen-1-ol,(E)-3-Hexenol,trans-1-Hydroxy-3-hexene,(3E)-Hexenol,trans-3-Hexenol IUPAC Name: (E)-hex-3-en-1-ol SMILES: CC\C=C\CCO